Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pgf
Entrez Gene ID
18654
Gene Name
placental growth factor
Gene Aliases
AI854365, PIGF, Plgf
Chromosome Location
Chr.12: 85166637 - 85177785 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271705.1 NP_001258634.1 2 - 3 126 58
NM_008827.3 NP_032853.1 2 - 3 459 58
GenBank mRNA AK042891.1 - 2 - 3 443 58
AK088943.1 - 2 - 3 442 58
BC016567.1 - 2 - 3 431 58
X80171.1 - 2 - 3 227 58
X96793.1 - 2 - 3 426 58

View Full Product Details