Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlh3
Entrez Gene ID
217716
Gene Name
mutL homolog 3 (E coli)
Gene Aliases
AV125803, BB126472
Chromosome Location
Chr.12: 85234466 - 85270648 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC003865.1 - 6 - 7 593 65

View Full Product Details