Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vash1
Entrez Gene ID
238328
Gene Name
vasohibin 1
Gene Aliases
AI834978, D930046M13Rik, G630009D10Rik
Chromosome Location
Chr.12: 86678700 - 86695688 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177354.4 NP_796328.2 2 - 3 1715 58
GenBank mRNA AB284948.1 - 2 - 3 1720 58
AK090166.1 - 2 - 3 1721 58
BC086688.1 - 2 - 3 215 58

View Full Product Details