The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199843.1 | NP_001186772.1 | 8 - 9 | 522 | 102 |
NM_029601.3 | NP_083877.1 | 7 - 8 | 489 | 102 | |
GenBank mRNA | AK006115.1 | - | 7 - 8 | 487 | 102 |
BC034668.1 | - | 8 - 9 | 460 | 102 | |
BC038915.1 | - | 8 - 9 | 511 | 102 | |
BE987571.1 | - | 1 - 2 | 102 | ||
CF751010.1 | - | 8 - 9 | 485 | 102 |