The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025888.5 | NP_080164.2 | 5 - 6 | 804 | 73 |
GenBank mRNA | AK010404.1 | - | 4 - 5 | 567 | 73 |
AK030202.1 | - | 5 - 6 | 806 | 73 | |
AK088807.1 | - | 4 - 5 | 572 | 73 | |
AK168440.1 | - | 5 - 6 | 795 | 73 | |
AK171742.1 | - | 4 - 5 | 602 | 73 | |
AK178692.1 | - | 1 - 2 | 56 | 73 | |
AK190659.1 | - | 1 - 2 | 56 | 73 | |
AK190821.1 | - | 1 - 2 | 56 | 73 | |
AK208777.1 | - | 1 - 2 | 87 | 73 | |
BC036318.1 | - | 3 - 4 | 449 | 73 | |
BC048161.1 | - | 4 - 5 | 588 | 73 | |
BC049250.1 | - | 4 - 5 | 681 | 73 | |
BC052712.1 | - | 4 - 5 | 590 | 73 |