The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009545.2 | NP_033571.1 | 2 - 3 | 350 | 78 |
GenBank mRNA | D90085.1 | - | 2 - 3 | 198 | 78 |