Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Med1
Entrez Gene ID
19014
Gene Name
mediator complex subunit 1
Gene Aliases
AI480703, CRSP210, DRIP205, PBP, Pparbp, TRAP220, l11Jus15
Chromosome Location
Chr.11: 98152154 - 98193314 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080118.1 NP_001073587.1 16 - 17 1703 114
NM_013634.2 NP_038662.2 16 - 17 1568 114
NM_134027.2 NP_598788.2 16 - 17 1703 114
GenBank mRNA AF000294.1 - 16 - 17 1568 114
AF332073.1 - 15 - 16 1448 114
AF332074.1 - 15 - 16 1448 114
AK049203.1 - 16 - 17 1694 114
AK147199.1 - 16 - 17 1680 114
BC021440.1 - 16 - 17 1674 114
BC079636.1 - 16 - 17 1673 114

View Full Product Details