Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pick1
Entrez Gene ID
18693
Gene Name
protein interacting with C kinase 1
Gene Aliases
Prkcabp
Chromosome Location
Chr.15: 79229166 - 79249466 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK050224.1 - 4 - 5 501 61

View Full Product Details