Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sbf1
Entrez Gene ID
77980
Gene Name
SET binding factor 1
Gene Aliases
2610510A08Rik, B230113C15Rik, Mtmr5, mKIAA3020
Chromosome Location
Chr.15: 89288236 - 89315311 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081030.2 NP_001074499.2 17 - 18 2165 70
NM_001170561.1 NP_001164032.1 17 - 18 2165 70
GenBank mRNA AK129485.1 - 17 - 18 2109 70
AK147332.1 - 17 - 18 2122 70
BC157935.1 - 17 - 18 2055 70
BC158013.1 - 17 - 18 2055 70
BC172094.1 - 17 - 18 2055 70

View Full Product Details