The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028722.2 | NP_082998.2 | 7 - 8 | 1752 | 132 |
GenBank mRNA | AK014463.1 | - | 7 - 8 | 2005 | 132 |
AK015496.1 | - | 6 - 7 | 968 | 132 | |
AK053719.1 | - | 6 - 7 | 1674 | 132 | |
BC058629.1 | - | 7 - 8 | 1752 | 132 |