The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177310.2 | NP_796284.1 | 8 - 9 | 1216 | 82 |
GenBank mRNA | AK088324.1 | - | 8 - 9 | 1216 | 82 |
BC137569.1 | - | 9 - 10 | 1378 | 82 | |
BC144717.1 | - | 9 - 10 | 1379 | 82 |