Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gngt2
Entrez Gene ID
14710
Gene Name
guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2
Gene Aliases
AV096488
Chromosome Location
Chr.11: 95842219 - 95845734 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038664.2 NP_001033753.2 1 - 2 128 92
GenBank mRNA AA028742.1 - 1 - 2 83 92
BC061005.1 - 1 - 2 78 92
W35863.1 - 1 - 2 137 92

View Full Product Details