Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpsg1
Entrez Gene ID
26945
Gene Name
tryptase gamma 1
Gene Aliases
Prss31, Tmt
Chromosome Location
Chr.17: 25369216 - 25374442 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK027982.1 - 1 - 2 148 62

View Full Product Details