Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Suz12
Entrez Gene ID
52615
Gene Name
suppressor of zeste 12 homolog (Drosophila)
Gene Aliases
2610028O16Rik, AI195385, AU016842, AW536442, D11Ertd530e, mKIAA0160
Chromosome Location
Chr.11: 79993106 - 80034123 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163018.1 NP_001156490.1 14 - 15 2022 85
NM_199196.2 NP_954666.1 15 - 16 2091 85
GenBank mRNA AK122213.1 - 15 - 16 2077 85
BC031375.1 - 1 - 2 41 85
BC039915.1 - 9 - 10 1167 85
BC051099.1 - 6 - 7 785 85
BC064461.1 - 15 - 16 2091 85
BC084591.1 - 10 - 11 1362 85

View Full Product Details