Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gsdma2
- Entrez Gene ID
-
76758
- Gene Name
- gasdermin A2
- Gene Aliases
-
2200001G21Rik, 2210006M16Rik, 2210009F20Rik, 2210411P14Rik, Gsdm2, Gsdml2
- Chromosome Location
-
Chr.11: 98646734 - 98657957 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK008685.1 |
- |
3 - 4 |
154 |
79 |
View Full Product Details