Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Utp6
Entrez Gene ID
216987
Gene Name
UTP6, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
4732497O03Rik, HCA66
Chromosome Location
Chr.11: 79932320 - 79962426 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144826.3 NP_659075.1 3 - 4 288 138
GenBank mRNA AF334387.1 - 3 - 4 615 138
AK029151.1 - 3 - 4 289 138
AK084976.1 - 3 - 4 281 138
AK164010.1 - 3 - 4 293 138
AK169785.1 - 3 - 4 295 138
BC018250.1 - 3 - 4 252 138
BC023920.1 - 3 - 4 282 138
BC024850.1 - 3 - 4 283 138

View Full Product Details