Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Suz12
Entrez Gene ID
52615
Gene Name
suppressor of zeste 12 homolog (Drosophila)
Gene Aliases
2610028O16Rik, AI195385, AU016842, AW536442, D11Ertd530e, mKIAA0160
Chromosome Location
Chr.11: 79993106 - 80034123 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199196.2 NP_954666.1 3 - 4 606 139
GenBank mRNA AK122213.1 - 3 - 4 591 139
AK207341.1 - 3 - 4 153 139
BC064461.1 - 3 - 4 606 139

View Full Product Details