Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slfn4
Entrez Gene ID
20558
Gene Name
schlafen 4
Gene Aliases
AI450778
Chromosome Location
Chr.11: 83175171 - 83190216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC046511.1 - 2 - 3 287 67

View Full Product Details