Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Actn1
Entrez Gene ID
109711
Gene Name
actinin, alpha 1
Gene Aliases
3110023F10Rik, Actn1a
Chromosome Location
Chr.12: 80167542 - 80260483 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ288940.1 - 19 - 20 2347 63

View Full Product Details