The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033537.2 | NP_001028709.2 | 3 - 4 | 528 | 63 |
GenBank mRNA | AK052486.1 | - | 3 - 4 | 445 | 63 |
AK078974.1 | - | 3 - 4 | 64 | ||
AK170817.1 | - | 3 - 4 | 443 | 63 | |
BC035945.1 | - | 3 - 4 | 451 | 63 |