The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053260.3 | NP_444490.2 | 5 - 6 | 838 | 69 |
GenBank mRNA | AF305425.2 | - | 4 - 5 | 674 | 69 |
AF442819.1 | - | 4 - 5 | 674 | 69 | |
AK144368.1 | - | 5 - 6 | 839 | 69 | |
BC104122.1 | - | 4 - 5 | 674 | 69 | |
BC104123.1 | - | 4 - 5 | 674 | 69 |