The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145968.2 | NP_666080.1 | 4 - 5 | 309 | 68 |
GenBank mRNA | BC030886.1 | - | 4 - 5 | 289 | 68 |
BC039780.1 | - | 4 - 5 | 300 | 68 | |
BC150728.1 | - | 4 - 5 | 254 | 68 |