Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim45
Entrez Gene ID
229644
Gene Name
tripartite motif-containing 45
Gene Aliases
4921530N01Rik
Chromosome Location
Chr.3: 100922493 - 100936925 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165953.1 NP_001159425.1 5 - 6 2009 81
GenBank mRNA AK076600.1 - 5 - 6 81

View Full Product Details