Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aldh6a1
Entrez Gene ID
104776
Gene Name
aldehyde dehydrogenase family 6, subfamily A1
Gene Aliases
1110038I05Rik, AI314632, Mmsdh
Chromosome Location
Chr.12: 84430717 - 84450950 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134042.2 NP_598803.1 9 - 10 1232 130
GenBank mRNA AF297860.1 - 9 - 10 1232 130
AK033587.1 - 9 - 10 1259 130
AK082790.1 - 9 - 10 1286 130
AK128924.1 - 9 - 10 1264 130
AK147146.1 - 9 - 10 1286 130
AK155814.1 - 9 - 10 1283 130
AK169915.1 - 9 - 10 1272 130
AK170305.1 - 9 - 10 1285 130
AK171581.1 - 9 - 10 1286 130
AK171896.1 - 9 - 10 1275 130
AK205140.1 - - 227 130
AK215618.1 - 1 - 2 103 130
AK219229.1 - 1 - 2 106 131
BC031148.1 - 9 - 10 1357 130
BC033440.1 - 8 - 9 1255 130

View Full Product Details