Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Car10
Entrez Gene ID
72605
Gene Name
carbonic anhydrase 10
Gene Aliases
2700029L05Rik, BB085816, Ca10
Chromosome Location
Chr.11: 93098142 - 93601751 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB080741.1 - 1 - 2 312 71
AK012302.1 - 1 - 2 319 71

View Full Product Details