The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161767.1 | NP_001155239.1 | 2 - 3 | 293 | 92 |
NM_001161768.1 | NP_001155240.1 | 2 - 3 | 290 | 92 | |
NM_172451.3 | NP_766039.2 | 2 - 3 | 289 | 92 | |
GenBank mRNA | AK028506.1 | - | 2 - 3 | 289 | 92 |
AK155008.1 | - | 2 - 3 | 296 | 92 | |
AK159721.1 | - | 2 - 3 | 271 | 92 |