Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spryd3
Entrez Gene ID
223918
Gene Name
SPRY domain containing 3
Gene Aliases
BC008150
Chromosome Location
Chr.15: 102116526 - 102136245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033277.3 NP_001028449.2 5 - 6 563 62
GenBank mRNA AK146689.1 - 5 - 6 566 62
BC008150.1 - 1 - 2 74 62
BC048820.1 - 4 - 5 792 62
BU057890.1 - 5 - 6 498 62
CX201967.1 - 1 - 2 62 62

View Full Product Details