Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spryd3
Entrez Gene ID
223918
Gene Name
SPRY domain containing 3
Gene Aliases
BC008150
Chromosome Location
Chr.15: 102116526 - 102136245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033277.3 NP_001028449.2 9 - 10 1078 87
GenBank mRNA AK146689.1 - 9 - 10 1081 87
BC008150.1 - 5 - 6 589 87
BC048820.1 - 8 - 9 1307 87
BG293312.1 - 2 - 3 163 87
CX201967.1 - 5 - 6 577 87

View Full Product Details