Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csde1
Entrez Gene ID
229663
Gene Name
cold shock domain containing E1, RNA binding
Gene Aliases
AA960392, BC016898, D3Jfr1, mKIAA0885, unr
Chromosome Location
Chr.3: 103020431 - 103058189 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161854.1 NP_001155326.1 2 - 3 546 113
NM_144901.3 NP_659150.1 2 - 3 546 113
GenBank mRNA AK044837.1 - 1 - 2 121 113
AK122393.1 - 2 - 3 456 113
AK144097.1 - 2 - 3 486 113
AK145351.1 - 2 - 3 651 113
AK145428.1 - 2 - 3 468 113
AK145932.1 - 2 - 3 463 113
AK146706.1 - 2 - 3 478 113
AK147012.1 - 2 - 3 485 113
AK150882.1 - 2 - 3 463 113
AK151638.1 - 2 - 3 482 113
AK151723.1 - 2 - 3 486 113
AK151970.1 - 2 - 3 486 113
AK152531.1 - 2 - 3 482 113
AK153196.1 - 2 - 3 463 113
AK154068.1 - 2 - 3 471 113
AK155751.1 - 2 - 3 471 113
AK159258.1 - 2 - 3 479 113
AK159863.1 - 2 - 3 479 113
BC016898.1 - 1 - 2 390 113
BC029688.1 - 2 - 3 482 113
BC031404.1 - 2 - 3 454 113
BC062097.1 - 2 - 3 461 113

View Full Product Details