Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eif2c5
Entrez Gene ID
286951
Gene Name
eukaryotic translation initiation factor 2C, 5
Gene Aliases
Ago5
Chromosome Location
Chr.X: 140776238 - 140796880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK090061.1 - 4 - 5 628 107
BC051684.1 - 2 - 3 196 107

View Full Product Details