Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nkiras2
- Entrez Gene ID
-
71966
- Gene Name
- NFKB inhibitor interacting Ras-like protein 2
- Gene Aliases
-
2410003M04Rik, 4930527H08Rik, D630018G21Rik, KBRAS2
- Chromosome Location
-
Chr.11: 100622955 - 100627602 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015918.1 |
- |
- |
59 |
59 |
View Full Product Details