Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cers5
- Entrez Gene ID
-
71949
- Gene Name
- ceramide synthase 5
- Gene Aliases
-
2310081H14Rik, AU045339, AW544927, Lass5, Trh4
- Chromosome Location
-
Chr.15: 99735592 - 99772515 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC043059.1 |
- |
- |
1115 |
79 |
View Full Product Details