Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsbn1
Entrez Gene ID
229675
Gene Name
rosbin, round spermatid basic protein 1
Gene Aliases
C230004D03Rik, E330004N01, Rosbin, Rsbn, Rsbp, mKIAA3002
Chromosome Location
Chr.3: 103914067 - 103966620 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172684.2 NP_766272.2 5 - 6 2157 151
GenBank mRNA AB101658.1 - 5 - 6 2159 151
AK087685.1 - 4 - 5 1123 151
AK122577.1 - 5 - 6 1692 151
AK134093.1 - 6 - 7 1276 151
AK138603.1 - 5 - 6 1701 151
BC094667.1 - 5 - 6 1682 151
BC139213.1 - 5 - 6 2126 151
BC139215.1 - 5 - 6 2126 151

View Full Product Details