Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm41
Entrez Gene ID
237073
Gene Name
RNA binding motif protein 41
Gene Aliases
BB131279, D330023I21Rik
Chromosome Location
Chr.X: 139889511 - 139998595 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK077972.1 - 1 - 2 60 107

View Full Product Details