The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081499.2 | NP_001074968.1 | 17 - 18 | 2907 | 89 |
GenBank mRNA | BC051701.1 | - | - | 873 | 89 |
BC147581.1 | - | 17 - 18 | 2770 | 89 | |
BC151057.1 | - | 17 - 18 | 2758 | 89 |