Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pacsin1
Entrez Gene ID
23969
Gene Name
protein kinase C and casein kinase substrate in neurons 1
Gene Aliases
A830061D09Rik, H74, mKIAA1379, syndapin
Chromosome Location
Chr.17: 27655591 - 27711118 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286743.1 NP_001273672.1 8 - 9 1350 126
NM_001286744.1 NP_001273673.1 8 - 9 1256 126
NM_011861.3 NP_035991.1 8 - 9 1315 126
NM_178365.4 NP_848142.1 8 - 9 1333 126
GenBank mRNA AK043967.1 - 8 - 9 1319 126
AK158686.1 - 8 - 9 1353 126
BC014698.1 - 8 - 9 1204 126
X85124.1 - 8 - 9 1224 126

View Full Product Details