Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Arl15
- Entrez Gene ID
-
218639
- Gene Name
- ADP-ribosylation factor-like 15
- Gene Aliases
-
A430036I03, Arfrp2, C230032K13Rik
- Chromosome Location
-
Chr.13: 113794506 - 114157462 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK053953.1 |
- |
1 - 2 |
103 |
104 |
View Full Product Details