The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023734.3 | NP_076223.3 | 5 - 6 | 1717 | 61 |
GenBank mRNA | AB046537.1 | - | - | 1366 | 61 |
BC039124.1 | - | 5 - 6 | 1314 | 61 |