Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stk38
Entrez Gene ID
106504
Gene Name
serine/threonine kinase 38
Gene Aliases
5830476G13Rik, 9530097A09Rik, AA617404, Ndr1
Chromosome Location
Chr.17: 28970885 - 29008006 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134115.2 NP_598876.1 13 - 14 1496 127
GenBank mRNA AY292399.1 - 13 - 14 1360 127
BC009658.1 - 13 - 14 1496 127

View Full Product Details