Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a13
Entrez Gene ID
239606
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 13
Gene Aliases
6530403A04, A630029G22Rik, AI505012, Gm308
Chromosome Location
Chr.15: 91267691 - 91573261 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033633.3 NP_001028805.2 8 - 9 1694 57
GenBank mRNA AK147341.1 - 8 - 9 1695 57
BC119586.1 - 9 - 10 1261 57
BC119587.1 - 8 - 9 1203 57

View Full Product Details