Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plxdc1
Entrez Gene ID
72324
Gene Name
plexin domain containing 1
Gene Aliases
2410003I07Rik, AI848450, Tem7
Chromosome Location
Chr.11: 97923237 - 97986667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163608.1 NP_001157080.1 11 - 12 1339 73
NM_028199.3 NP_082475.3 11 - 12 1318 73
GenBank mRNA AF378760.1 - 11 - 12 1252 73
AK010361.1 - 11 - 12 1318 73
AK036144.1 - 11 - 12 1293 73
AK153684.1 - 11 - 12 1305 73
BC114352.2 - 11 - 12 1238 73

View Full Product Details