The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163608.1 | NP_001157080.1 | 13 - 14 | 1540 | 94 |
NM_028199.3 | NP_082475.3 | 13 - 14 | 1519 | 94 | |
GenBank mRNA | AF378760.1 | - | 13 - 14 | 1453 | 94 |
AK010361.1 | - | 13 - 14 | 1519 | 94 | |
AK036144.1 | - | 13 - 14 | 1494 | 94 | |
AK153684.1 | - | 13 - 14 | 1506 | 94 | |
BC114352.2 | - | 13 - 14 | 1439 | 94 |