The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011945.2 | NP_036075.2 | 5 - 6 | 1150 | 82 |
GenBank mRNA | AB014614.1 | - | 5 - 6 | 1127 | 82 |
AF117340.1 | - | 5 - 6 | 1150 | 82 |