Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbp2
Entrez Gene ID
14120
Gene Name
fructose bisphosphatase 2
Gene Aliases
Fbp-1, Fbp1, Rae-30
Chromosome Location
Chr.13: 62836885 - 62858370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007994.3 NP_032020.2 2 - 3 404 68
GenBank mRNA AJ132692.1 - 2 - 3 331 68
AK146946.1 - 2 - 3 457 68
AK166898.1 - 2 - 3 405 68
AK166973.1 - 2 - 3 410 68
AK168374.1 - 2 - 3 435 68
BC012720.1 - 2 - 3 409 68
D42083.1 - 2 - 3 350 68

View Full Product Details