The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026072.1 | NP_080348.1 | 13 - 14 | 1471 | 84 |
GenBank mRNA | AK014025.1 | - | 13 - 14 | 1463 | 84 |
AK075902.1 | - | 13 - 14 | 1449 | 84 | |
AK077779.1 | - | 13 - 14 | 1414 | 84 | |
AK166770.1 | - | 13 - 14 | 1471 | 84 | |
BC025437.1 | - | 13 - 14 | 1271 | 84 |