Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cwc27
Entrez Gene ID
67285
Gene Name
CWC27 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases
3110009E13Rik, NY-CO-10, Sdccag10
Chromosome Location
Chr.13: 104631322 - 104816964 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026072.1 NP_080348.1 6 - 7 812 88
GenBank mRNA AK014025.1 - 6 - 7 795 88
AK075902.1 - 6 - 7 790 88
AK077779.1 - 6 - 7 755 88
AK166770.1 - 6 - 7 812 88

View Full Product Details