Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnh4
Entrez Gene ID
380728
Gene Name
potassium voltage-gated channel, subfamily H (eag-related), member 4
Gene Aliases
BEC2
Chromosome Location
Chr.11: 100740378 - 100760199 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081194.2 NP_001074663.1 16 - 17 3536 73

View Full Product Details