Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jup
Entrez Gene ID
16480
Gene Name
junction plakoglobin
Gene Aliases
Ctnng, D930025P04Rik, PG
Chromosome Location
Chr.11: 100368856 - 100397790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010593.2 NP_034723.1 12 - 13 2241 88
GenBank mRNA AK145895.1 - 12 - 13 2226 88
AK170934.1 - 12 - 13 2214 88
BC040757.1 - 12 - 13 2190 88
BC094461.1 - 12 - 13 2194 88
M90365.1 - 10 - 11 1706 88

View Full Product Details