Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxb4
Entrez Gene ID
15412
Gene Name
homeobox B4
Gene Aliases
Hox-2.6
Chromosome Location
Chr.11: 96318267 - 96321638 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010459.7 NP_034589.3 1 - 2 951 53
GenBank mRNA AK133945.1 - 1 - 2 480 53
BC095947.1 - 1 - 2 951 53
M36654.1 - 1 - 2 523 53

View Full Product Details