The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011862.3 | NP_035992.1 | 1 - 2 | 191 | 141 |
GenBank mRNA | AF128535.1 | - | 1 - 2 | 181 | 141 |
AK170299.1 | - | 1 - 2 | 150 | 141 | |
BY235086.1 | - | 1 - 2 | 192 | 141 |